missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HAI-2 Ectodomain Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
£168.00 - £487.00
Specifications
| Antigen | HAI-2/SPINT2 |
|---|---|
| Clone | 170018 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18707819
![]() |
R&D Systems
MAB1106-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18112681
|
R&D Systems
MAB1106 |
500 μg |
£487.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HAI-2/SPINT2 Monoclonal specifically detects HAI-2/SPINT2 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| HAI-2/SPINT2 | |
| LYOPH | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| O43291 | |
| 10653 | |
| Mouse myeloma cell line NS0-derived recombinant human HAI-2 aa 28-197 Accession # O43291 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects recombinant human (rh)HAI-2 in direct ELISAs and Western blots. In these formats, does not cross-react with rhHAI-1 or rmHAI-2B. |
| 170018 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HAI2, HAI2A, HAI2B, HAI-2DIAR3, HAI2FLJ45571, Hepatocyte growth factor activator inhibitor type 2, KOP, kunitz-type protease inhibitor 2, PB, Placental bikunin, serine peptidase inhibitor, Kunitz type, 2, serine protease inhibitor, Kunitz type, 2, SPINT2 | |
| SPINT2 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title