missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HAI-2 Ectodomain Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB1106-SP
This item is not returnable.
View return policy
Description
HAI-2/SPINT2 Monoclonal specifically detects HAI-2/SPINT2 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| HAI-2/SPINT2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| O43291 | |
| SPINT2 | |
| Mouse myeloma cell line NS0-derived recombinant human HAI-2 aa 28-197 Accession # O43291 | |
| 25 μg | |
| Primary | |
| Detects recombinant human (rh)HAI-2 in direct ELISAs and Western blots. In these formats, does not cross-react with rhHAI-1 or rmHAI-2B. | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 170018 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HAI2, HAI2A, HAI2B, HAI-2DIAR3, HAI2FLJ45571, Hepatocyte growth factor activator inhibitor type 2, KOP, kunitz-type protease inhibitor 2, PB, Placental bikunin, serine peptidase inhibitor, Kunitz type, 2, serine protease inhibitor, Kunitz type, 2, SPINT2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10653 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction