missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GRK3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£146.00 - £342.00
Specifications
| Antigen | GRK3 |
|---|---|
| Clone | 497007 |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 5-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18770349
![]() |
R&D Systems
MAB47851-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18752358
![]() |
R&D Systems
MAB47851-100 |
100 μg |
£342.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GRK3 Monoclonal specifically detects GRK3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
| GRK3 | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ADRBK2, adrenergic, beta, receptor kinase 2, BARK2, BARK2beta-adrenergic receptor kinase 2, beta-ARK-2, EC 2.7.11, EC 2.7.11.15, G-protein-coupled receptor kinase 3, GRK3Beta-ARK-2 | |
| E. coli-derived recombinant human GRK3 Pro469-Leu688 Accession # P35626 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human GRK3 in direct ELISAs. |
| 497007 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P35626 | |
| 157 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
RUO
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title