missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GRK3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB47851-100
This item is not returnable.
View return policy
Description
GRK3 Monoclonal specifically detects GRK3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications
| GRK3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ADRBK2, adrenergic, beta, receptor kinase 2, BARK2, BARK2beta-adrenergic receptor kinase 2, beta-ARK-2, EC 2.7.11, EC 2.7.11.15, G-protein-coupled receptor kinase 3, GRK3Beta-ARK-2 | |
| E. coli-derived recombinant human GRK3 Pro469-Leu688 Accession # P35626 | |
| 100 μg | |
| Primary | |
| Detects human GRK3 in direct ELISAs. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| 497007 | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 5-25 ug/mL | |
| P35626 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 157 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO
Spot an opportunity for improvement?Share a Content Correction