missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPR64 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£170.00 - £437.00
Specifications
| Antigen | GPR64 |
|---|---|
| Clone | 864238 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, CyTOF-ready |
| Applications | Flow Cytometry, Immunohistochemistry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18781097
![]() |
R&D Systems
MAB79771-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18740297
![]() |
R&D Systems
MAB79771-100 |
100 μg |
£437.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPR64 Monoclonal specifically detects GPR64 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
| GPR64 | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| EDDM6, epididymis-specific (seven transmembrane family), G protein-coupled receptor 64, HE6, MGC104454, MGC138738, TM7LN2 | |
| Chinese hamster ovary cell line CHO-derived recombinant human GPR64, Leu38-Asn64, Glu68-Thr553, Accession # NP_001073328 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 864238 | |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_001073328 | |
| 10149 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title