missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPR64 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB79771-SP
This item is not returnable.
View return policy
Description
GPR64 Monoclonal specifically detects GPR64 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Specifications
| GPR64 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| EDDM6, epididymis-specific (seven transmembrane family), G protein-coupled receptor 64, HE6, MGC104454, MGC138738, TM7LN2 | |
| Chinese hamster ovary cell line CHO-derived recombinant human GPR64, Leu38-Asn64, Glu68-Thr553, Accession # NP_001073328 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 864238 | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, CyTOF-ready | |
| NP_001073328 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10149 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction