missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human gp130 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 7 publications
£459.00 - £484.00
Specifications
| Antigen | gp130/CD130 |
|---|---|
| Concentration | LYOPH |
| Applications | Western Blot, Neutralization |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18043302
|
R&D Systems
AF-228-NA |
100 μg |
£484.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18032272
|
R&D Systems
AB-228-NA |
1 mg |
£459.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
gp130/CD130 Polyclonal specifically detects gp130/CD130 in Human samples. It is validated for Western Blot, Neutralization.Specifications
| gp130/CD130 | |
| Western Blot, Neutralization | |
| Unconjugated | |
| RUO | |
| P40189 | |
| 3572 | |
| IgG | |
| Affinity Purified |
| LYOPH | |
| Polyclonal | |
| Goat | |
| Human | |
| CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, IL6ST, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha | |
| IL6ST | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title