missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human gp130 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 7 publications
Brand: R&D Systems AF-228-NA
This item is not returnable.
View return policy
Description
gp130/CD130 Polyclonal specifically detects gp130/CD130 in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| gp130/CD130 | |
| Polyclonal | |
| Unconjugated | |
| P40189 | |
| IL6ST | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human gp130 Leu24-Glu619 (Glu619Asp) Accession # P40189 | |
| 100 μg | |
| Primary | |
| Detects human gp130 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant rat gp130, recombinant mouse gp130, recombinant human (rh) IL-1 sRI, rhIL-1 sRII, rhIL-2 sR beta , rhIL-2 sR gamma , rhIL-4 sR, and rhIL-6 sR is observed. | |
| Human | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 2-10 ug/mL | |
| CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, IL6ST, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3572 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction