missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GDNF Antibody, R&D Systems™

Mouse Monoclonal Antibody
£200.00 - £457.00
Specifications
| Antigen | GDNF |
|---|---|
| Clone | 27106R |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 μg;/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18792637
![]() |
R&D Systems
MAB212R-SP |
25 μg |
£200.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18712697
![]() |
R&D Systems
MAB212R-100 |
100 μg |
£457.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GDNF Monoclonal specifically detects GDNF in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| GDNF | |
| ELISA Capture (Matched Antibody Pair) 2-8 μg;/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P39905 | |
| 2668.0 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 27106R | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Astrocyte-derived trophic factor, ATF, ATF1, ATF2, glial cell derived neurotrophic factor, glial cell line derived neurotrophic factor, glial cell line-derived neurotrophic factor, HFB1-GDNF, hGDNF, HSCR3 | |
| E. coli-derived recombinant human GDNF, Arg109-Ile211, Accession No. P39905 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human GDNF in direct ELISA. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title