missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GDNF Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB212R-100
This item is not returnable.
View return policy
Description
GDNF Monoclonal specifically detects GDNF in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| GDNF | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Astrocyte-derived trophic factor, ATF, ATF1, ATF2, glial cell derived neurotrophic factor, glial cell line derived neurotrophic factor, glial cell line-derived neurotrophic factor, HFB1-GDNF, hGDNF, HSCR3 | |
| E. coli-derived recombinant human GDNF, Arg109-Ile211, Accession No. P39905 | |
| 100 μg | |
| 2668.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 27106R | |
| ELISA Capture (Matched Antibody Pair) 2-8 μg;/mL | |
| P39905 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human GDNF in direct ELISA. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction