missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Gas1 Antibody, R&D Systems™

Rat Monoclonal Antibody
£156.00 - £474.00
Specifications
| Antigen | Gas1 |
|---|---|
| Clone | 309325 |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18743857
![]() |
R&D Systems
MAB2636-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18140111
![]() |
R&D Systems
MAB2636 |
500 μg |
£474.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gas1 Monoclonal specifically detects Gas1 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| Gas1 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GAS-1, growth arrest-specific 1, Growth arrest-specific gene-1, growth arrest-specific protein 1 | |
| GAS1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 309325 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rat | |
| Human | |
| P54826 | |
| 2619 | |
| Mouse myeloma cell line NS0-derived recombinant human Gas1 Leu40-Ser318 Accession # P54826 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Gas1 in ELISAs and Western blots. In ELISAs, this antibody does not cross-react with recombinant mouse (rm) Gas1, recombinant human (rh) Gas6, rhArtemin, rhNeurturin, rhRet, or rhShh N-terminal domain. In Western blots, this antibody does not cross-react with rmGas6. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title