missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Gas1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2636-SP
This item is not returnable.
View return policy
Description
Gas1 Monoclonal specifically detects Gas1 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| Gas1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GAS-1, growth arrest-specific 1, Growth arrest-specific gene-1, growth arrest-specific protein 1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2619 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, ELISA | |
| 309325 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P54826 | |
| GAS1 | |
| Mouse myeloma cell line NS0-derived recombinant human Gas1 Leu40-Ser318 Accession # P54826 | |
| 25 μg | |
| Primary | |
| Detects human Gas1 in ELISAs and Western blots. In ELISAs, this antibody does not cross-react with recombinant mouse (rm) Gas1, recombinant human (rh) Gas6, rhArtemin, rhNeurturin, rhRet, or rhShh N-terminal domain. In Western blots, this antibody does not cross-react with rmGas6. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction