missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Galectin-8 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
£170.00 - £492.00
Specifications
| Antigen | Galectin-8 |
|---|---|
| Clone | 210608 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18769587
![]() |
R&D Systems
MAB1305-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18120901
|
R&D Systems
MAB1305 |
500 μg |
£492.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Galectin-8 Monoclonal specifically detects Galectin-8 in Human samples. It is validated for Western Blot.Specifications
| Galectin-8 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O00214 | |
| 3964 | |
| E. coli-derived recombinant human Galectin-8 Met1-Trp317 Accession # O00214 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Galectin-8 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Galectin-1, -2, or -7 is observed. |
| 210608 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Gal-8, GAL8, galectin 8, galectin-8, Galectin8, galectin-8g, lectin, galactoside-binding, soluble, 8, LGALS8, PCTA1, PCTA-1Po66 carbohydrate-binding protein, Po66 carbohydrate binding protein, Po66-CBP, Prostate carcinoma tumor antigen 1 | |
| LGALS8 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title