missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Galectin-8 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB1305-SP
This item is not returnable.
View return policy
Description
Galectin-8 Monoclonal specifically detects Galectin-8 in Human samples. It is validated for Western Blot.
Specifications
| Galectin-8 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O00214 | |
| LGALS8 | |
| E. coli-derived recombinant human Galectin-8 Met1-Trp317 Accession # O00214 | |
| 25 μg | |
| Primary | |
| Detects human Galectin-8 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Galectin-1, -2, or -7 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 210608 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Gal-8, GAL8, galectin 8, galectin-8, Galectin8, galectin-8g, lectin, galactoside-binding, soluble, 8, LGALS8, PCTA1, PCTA-1Po66 carbohydrate-binding protein, Po66 carbohydrate binding protein, Po66-CBP, Prostate carcinoma tumor antigen 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3964 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction