missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Galectin-12 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£146.00 - £396.00
Specifications
| Antigen | Galectin-12 |
|---|---|
| Clone | 984522 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18722497
![]() |
R&D Systems
MAB101571-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18762287
![]() |
R&D Systems
MAB101571-100 |
100 μg |
£396.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Galectin-12 Monoclonal specifically detects Galectin-12 in Human samples. It is validated for ELISA.Specifications
| Galectin-12 | |
| ELISA | |
| Monoclonal | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Gal-12, GAL12, galectin 12, Galectin-related inhibitor of proliferation, GRIP1, GRIP1galectin-12, lectin, galactoside-binding, soluble, 12, LGALS12 | |
| Human Galectin-12 synthetic peptide, Accession # Q96DT0 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Galectin-12 in direct ELISAs. |
| 984522 | |
| ELISA | |
| Unconjugated | |
| Human | |
| Q96DT0 | |
| 85329 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title