missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Galectin-12 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB101571-100
This item is not returnable.
View return policy
Description
Galectin-12 Monoclonal specifically detects Galectin-12 in Human samples. It is validated for ELISA.
Specifications
| Galectin-12 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Gal-12, GAL12, galectin 12, Galectin-related inhibitor of proliferation, GRIP1, GRIP1galectin-12, lectin, galactoside-binding, soluble, 12, LGALS12 | |
| Human Galectin-12 synthetic peptide, Accession # Q96DT0 | |
| 100 μg | |
| 85329 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| ELISA | |
| 984522 | |
| ELISA | |
| Q96DT0 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Galectin-12 in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction