missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human G-CSF Antibody, R&D Systems™

Mouse Monoclonal Antibody
£195.00 - £1750.00
Specifications
| Antigen | G-CSF |
|---|---|
| Clone | 3316R |
| Dilution | Neutralization 0.01-0.03 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Neutralization, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18734549
![]() |
R&D Systems
MAB214R-SP |
25 μg |
£195.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18727569
![]() |
R&D Systems
MAB214R-100 |
100 μg |
£425.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18171391
|
R&D Systems
MAB214R-01M |
1 mg |
£1460.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18199220
|
R&D Systems
MAB214R-500 |
500 ug |
£1750.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
G-CSF Monoclonal specifically detects G-CSF in Human samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| G-CSF | |
| Neutralization 0.01-0.03 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| NP_757373 | |
| 1440 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Detects human G-CSF in direct ELISAs. |
| 3316R | |
| Neutralization, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3, CSF3OS, Filgrastim, GCSF, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin | |
| CSF3 | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title