missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human G-CSF Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB214R-500
This item is not returnable.
View return policy
Description
G-CSF Monoclonal specifically detects G-CSF in Human samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| G-CSF | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. with No Preservative | |
| C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3, CSF3OS, Filgrastim, GCSF, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 1440 | |
| Human | |
| Purified |
| ELISA, Neutralization | |
| 3316R | |
| Neutralization 0.01-0.03 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| NP_757373 | |
| CSF3 | |
| E. coli-derived recombinant human G-CSF Ala30-Pro204 Accession No. NP_757373 | |
| 500 ug | |
| Primary | |
| Detects human G-CSF in direct ELISAs. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction