missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fucosyltransferase 3/5 (FUT3/5) Lumenal Domain Antibody, R&D Systems™

Sheep Polyclonal Antibody
£157.00 - £460.00
Specifications
| Antigen | Fucosyltransferase 3/5 (FUT3/5) |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18786857
![]() |
R&D Systems
AF4068-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18010122
![]() |
R&D Systems
AF4068 |
100 μg |
£460.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fucosyltransferase 3/5 (FUT3/5) Polyclonal specifically detects Fucosyltransferase 3/5 (FUT3/5) in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Fucosyltransferase 3/5 (FUT3/5) | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Sheep | |
| Human | |
| CD174, FT3B, FucT-III, FUC-TV, FUT3, FUT5, LE, Les, Lewis FT | |
| FUT3 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Q11128 | |
| 2525 | |
| Mouse myeloma cell line NS0-derived recombinant human FUT5 Arg35-Thr374 Accession # Q11128 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human FUT3 and human FUT5 in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title