missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fucosyltransferase 3/5 (FUT3/5) Lumenal Domain Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4068-SP
This item is not returnable.
View return policy
Description
Fucosyltransferase 3/5 (FUT3/5) Polyclonal specifically detects Fucosyltransferase 3/5 (FUT3/5) in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Fucosyltransferase 3/5 (FUT3/5) | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD174, FT3B, FucT-III, FUC-TV, FUT3, FUT5, LE, Les, Lewis FT | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 2525 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q11128 | |
| FUT3 | |
| Mouse myeloma cell line NS0-derived recombinant human FUT5 Arg35-Thr374 Accession # Q11128 | |
| 25 μg | |
| Primary | |
| Detects human FUT3 and human FUT5 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction