missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fucosyltransferase 3/5 (FUT3/5) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4068
This item is not returnable.
View return policy
Description
Fucosyltransferase 3/5 (FUT3/5) Monoclonal specifically detects Fucosyltransferase 3/5 (FUT3/5) in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Fucosyltransferase 3/5 (FUT3/5) | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| P21217 | |
| FUT3 | |
| Mouse myeloma cell line NS0-derived recombinant human Fucosyltransferase 3 Arg35-Thr361 Accession # P21217 | |
| 100 μg | |
| Primary | |
| Detects human Fucosyltransferase 3/5 (FUT3/5) in Western blots. The amino acid sequences of recombinant human FUT3 and recombinant human FUT5 are 91% identical. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 425037 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD174, FT3B, FucT-III, FUC-TV, FUT3, FUT5, LE, Les, Lewis FT | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2525 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction