missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fucosyltransferase 3/5 (FUT3/5) Antibody, R&D Systems™

Mouse Monoclonal Antibody
£157.00 - £352.00
Specifications
| Antigen | Fucosyltransferase 3/5 (FUT3/5) |
|---|---|
| Clone | 425037 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18758809
![]() |
R&D Systems
MAB4068-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18142651
![]() |
R&D Systems
MAB4068 |
100 μg |
£352.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fucosyltransferase 3/5 (FUT3/5) Monoclonal specifically detects Fucosyltransferase 3/5 (FUT3/5) in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Fucosyltransferase 3/5 (FUT3/5) | |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| P21217 | |
| 2525 | |
| Mouse myeloma cell line NS0-derived recombinant human Fucosyltransferase 3 Arg35-Thr361 Accession # P21217 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Fucosyltransferase 3/5 (FUT3/5) in Western blots. The amino acid sequences of recombinant human FUT3 and recombinant human FUT5 are 91% identical. |
| 425037 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD174, FT3B, FucT-III, FUC-TV, FUT3, FUT5, LE, Les, Lewis FT | |
| FUT3 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title