missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FSH beta Antibody, R&D Systems™

Rat Monoclonal Antibody
£157.00 - £369.00
Specifications
| Antigen | FSH beta |
|---|---|
| Clone | 961417 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18729777
![]() |
R&D Systems
MAB43102-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18748199
![]() |
R&D Systems
MAB43102-100 |
100 μg |
£369.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FSH beta Monoclonal specifically detects FSH beta in Human samples. It is validated for Western Blot.Specifications
| FSH beta | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Rat | |
| Human | |
| P01225 | |
| 2488 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 961417 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| follicle stimulating hormone, beta polypeptide, Follicle-stimulating hormone beta subunit, Follitropin, Follitropin beta chain, follitropin subunit beta, follitropin, beta chain, FSHB, FSH-B, FSH-beta | |
| E.coli-derived recombinant human FSH beta Asn19-Glu129 Accession # P01225 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human FSH beta in direct ELISAs and Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title