missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FSH beta Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB43102-100
This item is not returnable.
View return policy
Description
FSH beta Monoclonal specifically detects FSH beta in Human samples. It is validated for Western Blot.
Specifications
| FSH beta | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| follicle stimulating hormone, beta polypeptide, Follicle-stimulating hormone beta subunit, Follitropin, Follitropin beta chain, follitropin subunit beta, follitropin, beta chain, FSHB, FSH-B, FSH-beta | |
| E.coli-derived recombinant human FSH beta Asn19-Glu129 Accession # P01225 | |
| 100 μg | |
| Primary | |
| Detects human FSH beta in direct ELISAs and Western blots. | |
| Human | |
| IgG2b |
| Western Blot | |
| 961417 | |
| Western Blot 1 ug/mL | |
| P01225 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2488 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction