Learn More
Human FSH beta Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  R&D Systems MAB43102-SP
 View more versions of this product
Product Code. 18729777
  • £157.00 / 25µg
Description
FSH beta Monoclonal specifically detects FSH beta in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FSH beta
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
follicle stimulating hormone, beta polypeptide, Follicle-stimulating hormone beta subunit, Follitropin, Follitropin beta chain, follitropin subunit beta, follitropin, beta chain, FSHB, FSH-B, FSH-beta
E.coli-derived recombinant human FSH beta Asn19-Glu129 Accession # P01225
25 μg
Primary
Detects human FSH beta in direct ELISAs and Western blots.
Human
IgG2b
Western Blot
961417
Western Blot 1 ug/mL
P01225
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
2488
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.