missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGFR2 alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
£167.00 - £1765.00
Specifications
| Antigen | FGFR2 alpha |
|---|---|
| Clone | 98706 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18755119
![]() |
R&D Systems
MAB6841-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18756818
![]() |
R&D Systems
MAB6841-100 |
100 μg |
£454.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18736728
![]() |
R&D Systems
MAB6841-500 |
500 μg |
£1765.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGFR2 alpha Monoclonal specifically detects FGFR2 alpha in Human samples. It is validated for Western Blot.Specifications
| FGFR2 alpha | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| 2263 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human FGF R2 isoforms and Mouse myeloma cell line NS0-derived recombinant human FGF R2 isoforms | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human FGF R2 alpha in direct ELISAs and Western blots. Recognizes an epitope within the first Ig-like domain that is present only in alpha isoforms of human FGF R2. Does not cross-react with alpha isoforms of recombinant mouse FGF R2. |
| 98706 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FGFR2 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title