missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGFR2 alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB6841-SP
This item is not returnable.
View return policy
Description
FGFR2 alpha Monoclonal specifically detects FGFR2 alpha in Human samples. It is validated for Western Blot.
Specifications
| FGFR2 alpha | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 2263 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 98706 | |
| Western Blot 1 ug/mL | |
| FGFR2 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human FGF R2 isoforms and Mouse myeloma cell line NS0-derived recombinant human FGF R2 isoforms | |
| 25 μg | |
| Primary | |
| Detects human FGF R2 alpha in direct ELISAs and Western blots. Recognizes an epitope within the first Ig-like domain that is present only in alpha isoforms of human FGF R2. Does not cross-react with alpha isoforms of recombinant mouse FGF R2. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction