missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGF-19 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF969-SP
This item is not returnable.
View return policy
Beskrivning
FGF-19 Polyclonal specifically detects FGF-19 in Human samples. It is validated for Western Blot, Simple Western, Blockade of Receptor-ligand Interaction.
Specifikationer
| FGF-19 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FGF19, FGF-19, fibroblast growth factor 19 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9965 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 10 ug/mL, Blockade of Receptor-ligand Interaction | |
| O95750 | |
| FGF19 | |
| E. coli-derived recombinant human FGF-19 Phe27-Lys216 Accession # O95750 | |
| 25 μg | |
| Primary | |
| Detects human FGF-19 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) FGF-7, rhFGF-21 and recombinant mouse FGF-15 is observed. | |
| Human | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering