missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGF-19 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
£167.00 - £507.00
Specifications
| Antigen | FGF-19 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.5 ug/mL, Simple Western 10 ug/mL, Blockade of Receptor-ligand Interaction |
| Applications | Western Blot, Blockade of Receptor-ligand Interaction |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18774778
![]() |
R&D Systems
AF969-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18021002
|
R&D Systems
AF969 |
100 μg |
£507.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGF-19 Polyclonal specifically detects FGF-19 in Human samples. It is validated for Western Blot, Simple Western, Blockade of Receptor-ligand Interaction.Specifications
| FGF-19 | |
| Western Blot 0.5 ug/mL, Simple Western 10 ug/mL, Blockade of Receptor-ligand Interaction | |
| Polyclonal | |
| Goat | |
| Human | |
| FGF19, FGF-19, fibroblast growth factor 19 | |
| FGF19 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| Unconjugated | |
| RUO | |
| O95750 | |
| 9965 | |
| E. coli-derived recombinant human FGF-19 Phe27-Lys216 Accession # O95750 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human FGF-19 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) FGF-7, rhFGF-21 and recombinant mouse FGF-15 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title