missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGF-19 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF969
Additional Details : Weight : 0.00970kg
Description
FGF-19 Polyclonal specifically detects FGF-19 in Human samples. It is validated for Western Blot, Simple Western, Blockade of Receptor-ligand Interaction.Specifications
FGF-19 | |
Polyclonal | |
Unconjugated | |
O95750 | |
FGF19 | |
E. coli-derived recombinant human FGF-19 Phe27-Lys216 Accession # O95750 | |
100 μg | |
Primary | |
Detects human FGF-19 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) FGF-7, rhFGF-21 and recombinant mouse FGF-15 is observed. | |
Human | |
IgG |
Western Blot, Blockade of Receptor-ligand Interaction | |
LYOPH | |
Western Blot 0.5 ug/mL, Simple Western 10 ug/mL, Blockade of Receptor-ligand Interaction | |
FGF19, FGF-19, fibroblast growth factor 19 | |
Goat | |
Affinity Purified | |
RUO | |
9965 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only