missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGF-19 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£171.00 - £1675.00
Specifications
| Antigen | FGF-19 |
|---|---|
| Clone | 117611 |
| Dilution | Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, Blockade of Receptor-ligand Interaction, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18782789
![]() |
R&D Systems
MAB9691-SP |
25 μg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18756448
![]() |
R&D Systems
MAB9691-100 |
100 μg |
£417.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18743219
![]() |
R&D Systems
MAB9691-500 |
500 μg |
£1675.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGF-19 Monoclonal specifically detects FGF-19 in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair).Specifications
| FGF-19 | |
| Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| O95750 | |
| 9965 | |
| E. coli-derived recombinant human FGF-19 Phe27-Lys216 Accession # O95750 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human FGF-19 in direct ELISAs and Western blots. Shows approximately 10% cross-reactivity with recombinant human (rh) FGF-3 and rhFGF-5 and no cross-reactivity with rhFGF-4, rhFGF-6, rhFGF-7, rhFGF-9, rhFGF-10, rhFGF-13, rhFGF-16, rhFGF-17, rhFGF-18, rhFGF acidic, and rhFGF basic. |
| 117611 | |
| Western Blot, Blockade of Receptor-ligand Interaction, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FGF19, FGF-19, fibroblast growth factor 19 | |
| FGF19 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title