missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGF-19 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB9691-500
This item is not returnable.
View return policy
Description
FGF-19 Monoclonal specifically detects FGF-19 in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair).
Specifications
| FGF-19 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FGF19, FGF-19, fibroblast growth factor 19 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9965 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Blockade of Receptor-ligand Interaction, ELISA | |
| 117611 | |
| Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| O95750 | |
| FGF19 | |
| E. coli-derived recombinant human FGF-19 Phe27-Lys216 Accession # O95750 | |
| 500 μg | |
| Primary | |
| Detects human FGF-19 in direct ELISAs and Western blots. Shows approximately 10% cross-reactivity with recombinant human (rh) FGF-3 and rhFGF-5 and no cross-reactivity with rhFGF-4, rhFGF-6, rhFGF-7, rhFGF-9, rhFGF-10, rhFGF-13, rhFGF-16, rhFGF-17, rhFGF-18, rhFGF acidic, and rhFGF basic. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction