missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FAM3B Antibody, R&D Systems™

Mouse Monoclonal Antibody
£157.00 - £352.00
Specifications
| Antigen | FAM3B |
|---|---|
| Clone | 365505 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18723798
![]() |
R&D Systems
MAB2866-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18153321
![]() |
R&D Systems
MAB2866 |
100 μg |
£352.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FAM3B Monoclonal specifically detects FAM3B in Human samples. It is validated for Western Blot.Specifications
| FAM3B | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_478066 | |
| 54097 | |
| Mouse myeloma cell line NS0-derived recombinant human FAM3B isoform B Glu30-Ser235 Accession # NP_478066 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human FAM3B in direct ELISAs and Western blots. In direct ELISAs and Western blots, 75-100% cross-reactivity with recombinant mouse FAM3B and no cross-reactivity with recombinant human (rh) FAM3A, rhFAM3C, or rhFAM3D is observed. |
| 365505 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 2-21, C21orf11, C21orf76PANDER, chromosome 21 open reading frame 11, Cytokine-like protein 2-21, D21M16SJHU19e, family with sequence similarity 3, member B, ORF9, pancreatic derived factor, Pancreatic-derived factor, PANDER, PRED44 | |
| FAM3B | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title