missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FAM3B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2866-SP
This item is not returnable.
View return policy
Description
FAM3B Monoclonal specifically detects FAM3B in Human samples. It is validated for Western Blot.
Specifications
| FAM3B | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| NP_478066 | |
| FAM3B | |
| Mouse myeloma cell line NS0-derived recombinant human FAM3B isoform B Glu30-Ser235 Accession # NP_478066 | |
| 25 μg | |
| Primary | |
| Detects human FAM3B in direct ELISAs and Western blots. In direct ELISAs and Western blots, 75-100% cross-reactivity with recombinant mouse FAM3B and no cross-reactivity with recombinant human (rh) FAM3A, rhFAM3C, or rhFAM3D is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 365505 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 2-21, C21orf11, C21orf76PANDER, chromosome 21 open reading frame 11, Cytokine-like protein 2-21, D21M16SJHU19e, family with sequence similarity 3, member B, ORF9, pancreatic derived factor, Pancreatic-derived factor, PANDER, PRED44 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 54097 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction