Learn More
Human Erythropoietin/EPO Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB2871-500
 View more versions of this product
Product Code. 18730927
  • £1915.00 / 500µg
Description
Erythropoietin/EPO Monoclonal specifically detects Erythropoietin/EPO in Human samples. It is validated for Western Blot, Immunoaffinity Purification.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Erythropoietin/EPO
Monoclonal
Unconjugated
Supplied as a 0.2 μm filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
2056
Human
Supernatant
Western Blot, Affinity Purification
AE7A5
Western Blot 1 ug/mL, Immunoaffinity Purification
P01588
EPO
Erythropoietin/EPO synthetic peptide analog, Ala28-Thr53, Accession # P01588
500 μg
Primary
Recognizes an epitope within the first 26 amino acids at the NH2 terminus of human urinary and human recombinant EPO. Binds human and rat EPO but does not bind sheep, baboon, or dog EPO (1, 2).
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening.
IgG2a