missing translation for 'onlineSavingsMsg'
Learn More

Human Erythropoietin/EPO Antibody, R&D Systems™

Product Code. p-5380166
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
Unit Size:
100µg
25µg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18762477 25 μg 25µg
18741127 100 μg 100µg
18730927 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18762477 Supplier R&D Systems Supplier No. MAB2871SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Erythropoietin/EPO Monoclonal specifically detects Erythropoietin/EPO in Human samples. It is validated for Western Blot, Immunoaffinity Purification.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Erythropoietin/EPO
Applications Western Blot, Affinity Purification
Classification Monoclonal
Clone AE7A5
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoaffinity Purification
Formulation Supplied as a 0.2 μm filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P01588
Gene Alias ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2
Gene Symbols EPO
Host Species Mouse
Immunogen Erythropoietin/EPO synthetic peptide analog, Ala28-Thr53, Accession # P01588
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2056
Test Specificity Recognizes an epitope within the first 26 amino acids at the NH2 terminus of human urinary and human recombinant EPO. Binds human and rat EPO but does not bind sheep, baboon, or dog EPO (1, 2).
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.