missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Eps15 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£178.00 - £409.00
Specifications
| Antigen | EPS15 |
|---|---|
| Clone | 1261C |
| Dilution | Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 1-25 ug/mL |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18715209
![]() |
R&D Systems
MAB8480-SP |
25 μg |
£178.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18741319
![]() |
R&D Systems
MAB8480 |
100 μg |
£409.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EPS15 Monoclonal specifically detects EPS15 in Human samples. It is validated for Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, Immunocytochemistry.Specifications
| EPS15 | |
| Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 1-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| AF1P, AF-1P, ALL1 fused gene from chromosome 1, epidermal growth factor receptor pathway substrate 15, epidermal growth factor receptor substrate 15, MLLT5, Protein AF-1p, Protein Eps15 | |
| EPS15 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 1261C | |
| Western Blot, Flow Cytometry, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P42566 | |
| 2060 | |
| E.coli-derived recombinant human Eps15 Ala448-Thr579 Accession # P42566 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS | |
| Detects human Eps15 in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title