missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Eps15 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB8480-SP
This item is not returnable.
View return policy
Description
EPS15 Monoclonal specifically detects EPS15 in Human samples. It is validated for Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, Immunocytochemistry.
Specifications
| EPS15 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| AF1P, AF-1P, ALL1 fused gene from chromosome 1, epidermal growth factor receptor pathway substrate 15, epidermal growth factor receptor substrate 15, MLLT5, Protein AF-1p, Protein Eps15 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 2060 | |
| Reconstitute at 0.5 mg/mL in sterile PBS | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Flow Cytometry, Immunocytochemistry | |
| 1261C | |
| Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 1-25 ug/mL | |
| P42566 | |
| EPS15 | |
| E.coli-derived recombinant human Eps15 Ala448-Thr579 Accession # P42566 | |
| 25 μg | |
| Primary | |
| Detects human Eps15 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction