missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Epimorphin/Syntaxin 2 Antibody, R&D Systems™

Goat Polyclonal Antibody
£167.00 - £441.00
Specifications
| Antigen | Epimorphin/Syntaxin 2 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18717777
![]() |
R&D Systems
AF2936-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18079371
|
R&D Systems
AF2936 |
100 μg |
£441.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Epimorphin/Syntaxin 2 Polyclonal specifically detects Epimorphin/Syntaxin 2 in Human samples. It is validated for Western Blot.Specifications
| Epimorphin/Syntaxin 2 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| EPIMepimorphin, EPM, STX2, STX2AMGC51014, STX2Bsyntaxin-2, STX2CEpimorphin, Syntaxin 2 | |
| STX2 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| P32856 | |
| 2054 | |
| E. coli-derived recombinant human Epimorphin/Syntaxin 2 Met1-Arg188 Accession # P32856 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Epimorphin/Syntaxin 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 60% cross-reactivity with recombinant mouse Epimorphin is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title