missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Epimorphin/Syntaxin 2 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF2936
This item is not returnable.
View return policy
Description
Epimorphin/Syntaxin 2 Polyclonal specifically detects Epimorphin/Syntaxin 2 in Human samples. It is validated for Western Blot.
Specifications
| Epimorphin/Syntaxin 2 | |
| Polyclonal | |
| Unconjugated | |
| P32856 | |
| STX2 | |
| E. coli-derived recombinant human Epimorphin/Syntaxin 2 Met1-Arg188 Accession # P32856 | |
| 100 μg | |
| Primary | |
| Detects human Epimorphin/Syntaxin 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 60% cross-reactivity with recombinant mouse Epimorphin is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| EPIMepimorphin, EPM, STX2, STX2AMGC51014, STX2Bsyntaxin-2, STX2CEpimorphin, Syntaxin 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2054 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction