missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EphA1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£168.00 - £487.00
Specifications
| Antigen | EphA1 |
|---|---|
| Clone | 86810 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18757219
![]() |
R&D Systems
MAB638-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18141761
|
R&D Systems
MAB638 |
500 μg |
£487.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EphA1 Monoclonal specifically detects EphA1 in Human samples. It is validated for Western Blot.Specifications
| EphA1 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.10, EC 2.7.10.1, EPH receptor A1, eph tyrosine kinase 1, EphA1, EPHephrin type-A receptor 1, EPHT, EPHT1erythropoietin-producing hepatoma amplified sequence, Esk, MGC163163, oncogene EPH, soluble EPHA1 variant 1, soluble EPHA1 variant 2, Tyrosine-protein kinase receptor EPH | |
| EPHA1 | |
| IgG2a | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 86810 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P21709 | |
| 2041 | |
| Mouse myeloma cell line NS0-derived recombinant human EphA1 Arg24-Glu547 (predicted) Accession # P21709 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human EphA1 in direct ELISAs and Western blots. No cross-reactivity with recombinant mouse EphA2, A3, A4, A6, A7, A8, B2, B4, B5, B6, recombinant rat (rr) EphA5, rrEphB1, or recombinant human EphB1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title