missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EphA1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB638
This item is not returnable.
View return policy
Description
EphA1 Monoclonal specifically detects EphA1 in Human samples. It is validated for Western Blot.
Specifications
| EphA1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.10, EC 2.7.10.1, EPH receptor A1, eph tyrosine kinase 1, EphA1, EPHephrin type-A receptor 1, EPHT, EPHT1erythropoietin-producing hepatoma amplified sequence, Esk, MGC163163, oncogene EPH, soluble EPHA1 variant 1, soluble EPHA1 variant 2, Tyrosine-protein kinase receptor EPH | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 2041 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 86810 | |
| Western Blot 1 ug/mL | |
| P21709 | |
| EPHA1 | |
| Mouse myeloma cell line NS0-derived recombinant human EphA1 Arg24-Glu547 (predicted) Accession # P21709 | |
| 500 μg | |
| Primary | |
| Detects human EphA1 in direct ELISAs and Western blots. No cross-reactivity with recombinant mouse EphA2, A3, A4, A6, A7, A8, B2, B4, B5, B6, recombinant rat (rr) EphA5, rrEphB1, or recombinant human EphB1 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction