missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DPP9 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £377.00
Specifications
| Antigen | DPP9 |
|---|---|
| Clone | 988515 |
| Dilution | Western Blot 1μg/mL, Simple Western 10μg/mL, Immunocytochemistry 8-25μg/mL |
| Applications | Immunocytochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18181221
![]() |
R&D Systems
MAB54191-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18140531
![]() |
R&D Systems
MAB54191-100 |
100 μg |
£377.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
DPP9 Monoclonal specifically detects DPP9 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Tekniske data
| DPP9 | |
| Western Blot 1μg/mL, Simple Western 10μg/mL, Immunocytochemistry 8-25μg/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| Q86TI2 | |
| 91039 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 988515 | |
| Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| dipeptidyl peptidase 9, Dipeptidyl peptidase IV-related protein 2, dipeptidyl peptidase IV-related protein-2, Dipeptidyl peptidase IX, Dipeptidyl peptidase-like protein 9, dipeptidylpeptidase 9, dipeptidyl-peptidase 9, DKFZp762F117, DP9, DPLP9, DPP IX, DPRP2, DPRP-2, EC 3.4.14.5, FLJ16073 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human DPP9 Arg2-Leu892 Accession # Q86TI2 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human DPP9 in ELISAs. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel