missing translation for 'onlineSavingsMsg'
Learn More

Human DPP9 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18181221
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18181221 25 μg 25µg
18140531 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18181221 Supplier R&D Systems Supplier No. MAB54191SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DPP9 Monoclonal specifically detects DPP9 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPP9
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 988515
Conjugate Unconjugated
Dilution Western Blot 1μg/mL, Simple Western 10μg/mL, Immunocytochemistry 8-25μg/mL
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gene Accession No. Q86TI2
Gene Alias dipeptidyl peptidase 9, Dipeptidyl peptidase IV-related protein 2, dipeptidyl peptidase IV-related protein-2, Dipeptidyl peptidase IX, Dipeptidyl peptidase-like protein 9, dipeptidylpeptidase 9, dipeptidyl-peptidase 9, DKFZp762F117, DP9, DPLP9, DPP IX, DPRP2, DPRP-2, EC 3.4.14.5, FLJ16073
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human DPP9 Arg2-Leu892 Accession # Q86TI2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 91039
Test Specificity Detects human DPP9 in ELISAs.
Reconstitution Reconstitute at 0.5mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.