missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DNAM-1/CD226 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
£170.00 - £1810.00
Specifications
| Antigen | DNAM-1/CD226 |
|---|---|
| Clone | 102511 |
| Dilution | Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Neutralization 0.1-0.6 ug/mL, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Neutralization, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18780929
![]() |
R&D Systems
MAB666-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18128980
![]() |
R&D Systems
MAB666-100 |
100 μg |
£448.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18713857
![]() |
R&D Systems
MAB666-500 |
500 μg |
£1810.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNAM-1/CD226 Monoclonal specifically detects DNAM-1/CD226 in Human samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOF-ready.Specifications
| DNAM-1/CD226 | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Neutralization 0.1-0.6 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q15762 | |
| 10666 | |
| Mouse myeloma cell line NS0-derived recombinant human DNAM-1/CD226 Glu19-Asn247 (predicted) Accession # Q15762 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human DNAM-1/CD226 in direct ELISAs and Western blots. |
| 102511 | |
| Western Blot, Flow Cytometry, Neutralization, CyTOF | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD226, CD226 antigenplatelet and T cell activation antigen 1, CD226 molecule, DNAM1, DNAM1adhesion glycoprotein, DNAM-1DNAX accessory molecule-1, DNAX accessory molecule 1, PTA1, T lineage-specific activation antigen 1 antigen, TLiSA1 | |
| CD226 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title