Learn More
Human DNAM-1/CD226 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 6 publications
Brand:  R&D Systems MAB666-500
 View more versions of this product
Product Code. 18713857
  • £1810.00 / 500µg
Description
DNAM-1/CD226 Monoclonal specifically detects DNAM-1/CD226 in Human samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DNAM-1/CD226
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD226, CD226 antigenplatelet and T cell activation antigen 1, CD226 molecule, DNAM1, DNAM1adhesion glycoprotein, DNAM-1DNAX accessory molecule-1, DNAX accessory molecule 1, PTA1, T lineage-specific activation antigen 1 antigen, TLiSA1
Mouse
Protein A or G purified from ascites
RUO
10666
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1
Western Blot, Flow Cytometry, Neutralization, CyTOF
102511
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Neutralization 0.1-0.6 ug/mL, CyTOF-ready
Q15762
CD226
Mouse myeloma cell line NS0-derived recombinant human DNAM-1/CD226 Glu19-Asn247 (predicted) Accession # Q15762
500 μg
Primary
Detects human DNAM-1/CD226 in direct ELISAs and Western blots.
Human
Ascites