missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Dkk-4 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB1269-050
Additional Details : Weight : 0.00970kg
Description
Dkk-4 Monoclonal specifically detects Dkk-4 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
Dkk-4 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Q9UBT3 | |
DKK4 | |
Mouse myeloma cell line NS0-derived recombinant human Dkk-4 Leu19-Leu224 Accession # Q9UBT3 | |
50 μg | |
Primary | |
Detects human Dkk-4 in ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant mouse (rm) Dkk-4 is observed and no cross-reactivity with recombinant human (rh) Dkk-1 or rhDkk-3 is observed. In sandwich immunoassays, less than 2% cross-reactivity with rmDkk-4 is observed and no cross-reactivity with rhDkk-1, recombinant rat Dkk-1, rmDkk-1, or rhDkk-3 is observed. | |
Human | |
Purified |
Western Blot, ELISA | |
185709 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
dickkopf (Xenopus laevis) homolog 4, dickkopf homolog 4 (Xenopus laevis), dickkopf-4, dickkopf-related protein 4, Dkk4, DKK-4, hDkk-4, MGC129562, MGC129563 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
27121 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |