missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Dkk-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£168.00 - £1520.00
Specifications
| Antigen | Dkk-4 |
|---|---|
| Clone | 185709 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18787049
![]() |
R&D Systems
MAB1269-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18123431
![]() |
R&D Systems
MAB1269-050 |
50 μg |
£254.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18130531
![]() |
R&D Systems
MAB1269-500 |
500 μg |
£1520.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dkk-4 Monoclonal specifically detects Dkk-4 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| Dkk-4 | |
| LYOPH | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9UBT3 | |
| 27121 | |
| Mouse myeloma cell line NS0-derived recombinant human Dkk-4 Leu19-Leu224 Accession # Q9UBT3 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Dkk-4 in ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant mouse (rm) Dkk-4 is observed and no cross-reactivity with recombinant human (rh) Dkk-1 or rhDkk-3 is observed. In sandwich immunoassays, less than 2% cross-reactivity with rmDkk-4 is observed and no cross-reactivity with rhDkk-1, recombinant rat Dkk-1, rmDkk-1, or rhDkk-3 is observed. |
| 185709 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| dickkopf (Xenopus laevis) homolog 4, dickkopf homolog 4 (Xenopus laevis), dickkopf-4, dickkopf-related protein 4, Dkk4, DKK-4, hDkk-4, MGC129562, MGC129563 | |
| DKK4 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title