missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Dectin-1/CLEC7A Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 6 publications
£180.00 - £511.00
Specifications
| Antigen | Dectin-1/CLEC7A |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction |
| Applications | Western Blot, Blockade of Receptor-ligand Interaction |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18786107
![]() |
R&D Systems
AF1859-SP |
25 μg |
£180.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18051352
![]() |
R&D Systems
AF1859 |
100 μg |
£511.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dectin-1/CLEC7A Polyclonal specifically detects Dectin-1/CLEC7A in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.Specifications
| Dectin-1/CLEC7A | |
| Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction | |
| Polyclonal | |
| Goat | |
| Human | |
| Beta-glucan receptor, BGR, CD369, CLEC7A, CLECSF12, CLECSF12DC-associated C-type lectin 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 12, C-type lectin domain family 7 member A, C-type lectin domain family 7, member A, C-type lectin superfamily member 12, dectin-1, Dectin1, DECTIN1CANDF4, Dendritic cell-associated C-type lectin 1, dendritic cell-associated C-type lectin-1, hDectin-1, lectin-like receptor 1 | |
| CLEC7A | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| Unconjugated | |
| RUO | |
| NP_072092 | |
| 64581 | |
| Mouse myeloma cell line NS0-derived recombinant human Dectin-1/CLEC7A (R&D Systems, Catalog # 1859-DC) Thr66-Met201 Accession # NP_072092 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Dectin-1/CLEC7A in direct ELISAs and Western blots. In this format, less than 2% cross-reactivity with recombinant mouse Dectin-1 and recombinant human Dectin-2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title