missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Dectin-1/CLEC7A Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 6 publications
Brand: R&D Systems AF1859
This item is not returnable.
View return policy
Description
Dectin-1/CLEC7A Polyclonal specifically detects Dectin-1/CLEC7A in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.
Specifications
| Dectin-1/CLEC7A | |
| Polyclonal | |
| Unconjugated | |
| NP_072092 | |
| CLEC7A | |
| Mouse myeloma cell line NS0-derived recombinant human Dectin-1/CLEC7A (R&D Systems, Catalog # 1859-DC) Thr66-Met201 Accession # NP_072092 | |
| 100 μg | |
| Primary | |
| Detects human Dectin-1/CLEC7A in direct ELISAs and Western blots. In this format, less than 2% cross-reactivity with recombinant mouse Dectin-1 and recombinant human Dectin-2 is observed. | |
| Human | |
| IgG |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction | |
| Beta-glucan receptor, BGR, CD369, CLEC7A, CLECSF12, CLECSF12DC-associated C-type lectin 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 12, C-type lectin domain family 7 member A, C-type lectin domain family 7, member A, C-type lectin superfamily member 12, dectin-1, Dectin1, DECTIN1CANDF4, Dendritic cell-associated C-type lectin 1, dendritic cell-associated C-type lectin-1, hDectin-1, lectin-like receptor 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 64581 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction